Please login to your online account to display your discounted pricing

Aluminium ammonium sulfate dodecahydrate, 99%, pure, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  206260050

 View more versions of this product

Catalog No. AC206260050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 453.33
Physical Form Crystals
Quantity 5g
Color White
Packaging Glass Bottle
Purity Grade Pure
Density 1.6400g/mL
Chemical Name or Material Aluminium ammonium sulfate dodecahydrate
MDL Number MFCD00149958
CAS 7784-26-1
Loss on Drying 45% to 48% (2 g, 300°C)
Linear Formula AlNH4(SO4)2·12H2O
Synonym Ammonium alum
Molecular Formula H4AlNO8S2·12H2O
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Aluminium ammonium sulfate dodecahydrate
CAS 7784-26-1
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula H4AlNO8S2·12H2O
Linear Formula AlNH4(SO4)2·12H2O
Formula Weight 453.33
MDL Number MFCD00149958
Synonym Ammonium alum
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents