missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Aluminium acetylacetonate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC196975000

 View more versions of this product

Catalog No. AC196975000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Aluminium acetylacetonate
96.0
Authentic
Crystalline Powder
97%
(CH3COCH2COCH2)3Al
MFCD00000013
White to cream
500g
13963-57-0
100.0
Plastic bottle
≥96.0%
C15H21AlO6
324.3
Aluminium 2, 4-pentanedionate
190° to 195°C

Chemical Identifiers

13963-57-0
100.0
C15H21AlO6
324.3
Aluminium 2, 4-pentanedionate
≥96.0%
96.0
97%
(CH3COCH2COCH2)3Al
MFCD00000013
Aluminium acetylacetonate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger