Please login to your online account to display your discounted pricing

Aluminium acetylacetonate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  196970050

 View more versions of this product

Catalog No. AC196970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 324.3
Quantity 5g
Color White to cream
Melting Point 190° to 195°C
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C15H21AlO6
MDL Number MFCD00000013
CAS 13963-57-0
Chemical Name or Material Aluminium acetylacetonate
Infrared Spectrum Authentic
Linear Formula (CH3COCH2COCH2)3Al
Synonym Aluminium 2, 4-pentanedionate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.

Specifications

Chemical Name or Material Aluminium acetylacetonate
CAS 13963-57-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C15H21AlO6
Linear Formula (CH3COCH2COCH2)3Al
Formula Weight 324.3
MDL Number MFCD00000013
Synonym Aluminium 2, 4-pentanedionate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.

SDS & Safety Documents