Please login to your online account to display your discounted pricing

Aluminium, 99%, powder, 200 mesh, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  200935000

 View more versions of this product

Catalog No. AC200935000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Finely Divided Powder
Formula Weight 26.98
Quantity 500g
Color Gray
Merck Index 15, 319
EINECS Number 231-072-3
TSCA TSCA
Density 2.7
Particle Size 75 Micron (>95% passes sieve)
RTECS Number BD0330000
Fieser 02,19; 04,9; 12,486; 16,337
Molecular Formula Al
CAS 7429-90-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Trace Analysis 800ppm max.
Chemical Name or Material Aluminium
Copper (Cu) 300ppm max.
Name Note 200 mesh, 99%
Assay Percent Range 99%
Iron (Fe) 6000ppm max.
MDL Number MFCD00134029
Packaging Plastic bottle
Silicon (Si) 5000ppm max.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Aluminium
Name Note 200 mesh, 99%
CAS 7429-90-5
Assay Percent Range 99%
Molecular Formula Al
Formula Weight 26.98
MDL Number MFCD00134029
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.

SDS & Safety Documents