Please login to your online account to display your discounted pricing

Aluminium, 99%, powder, 200 mesh, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  200930025

 View more versions of this product

Catalog No. AC200930025

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 26.98
Physical Form Finely Divided Powder
Quantity 2.5kg
Color Gray
Merck Index 15, 319
TSCA TSCA
EINECS Number 231-072-3
RTECS Number BD0330000
Density 2.7
Particle Size 75 Micron (>95% passes sieve)
Fieser 02,19; 04,9; 12,486; 16,337
Silicon (Si) 5000ppm max.
Iron (Fe) 6000ppm max.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 200 mesh, 99%
MDL Number MFCD00134029
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Molecular Formula Al
Chemical Name or Material Aluminium
CAS 7429-90-5
Packaging Plastic bottle
Copper (Cu) 300ppm max.
Trace Analysis 800ppm max.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Aluminium
Name Note 200 mesh, 99%
CAS 7429-90-5
Assay Percent Range 99%
Molecular Formula Al
Formula Weight 26.98
MDL Number MFCD00134029
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.

SDS & Safety Documents