Please login to your online account to display your discounted pricing

Aluminium, 99%, powder, 200 mesh, ACROS Organics™

$50.60 - $278.50

Chemical Identifiers

Chemical Name or Material Aluminium
Name Note 200 mesh, 99%
CAS 7429-90-5
Assay Percent Range 99%
Molecular Formula Al
Formula Weight 26.98
MDL Number MFCD00134029
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC200930025

  SDS
thermo fisher scientific
200930025
Plastic bottle 2.5kg Each for $278.50

AC200930250

  SDS
thermo fisher scientific
200930250
Plastic bottle 25g Each for $50.60

AC200935000

  SDS
fisher scientific acros
200935000
Plastic bottle 500g Each for $82.39
Description & Specifications

Specifications

Formula Weight 26.98
Physical Form Finely Divided Powder
Quantity 2.5kg
Color Gray
Merck Index 15, 319
TSCA TSCA
EINECS Number 231-072-3
RTECS Number BD0330000
Density 2.7
Particle Size 75 Micron (>95% passes sieve)
Fieser 02,19; 04,9; 12,486; 16,337
Silicon (Si) 5000ppm max.
Iron (Fe) 6000ppm max.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 200 mesh, 99%
MDL Number MFCD00134029
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Molecular Formula Al
Chemical Name or Material Aluminium
CAS 7429-90-5
Packaging Plastic bottle
Copper (Cu) 300ppm max.
Trace Analysis 800ppm max.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.