Please login to your online account to display your discounted pricing

Aluminium, 99.5%, shot (2-10 mm), ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  300212500

 View more versions of this product

Catalog No. AC300212500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Shot
Formula Weight 26.97
Quantity 250g
Color Gray
EINECS Number 231-072-3
TSCA TSCA
Density 2.7000g/mL
RTECS Number BD0330000
Fieser 02,19; 04,9; 12,486; 16,337
Chemical Name or Material Aluminium
Name Note 99.5%
Assay Percent Range 99.5%
Molecular Formula Al
MDL Number MFCD00134029
CAS 7429-90-5
Packaging Glass bottle
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 AIR SENSITIVE
Total Trace Metal Impurities 0.5% max.

Specifications

Chemical Name or Material Aluminium
Name Note 99.5%
CAS 7429-90-5
Assay Percent Range 99.5%
Molecular Formula Al
Formula Weight 26.97
MDL Number MFCD00134029
Accessories

MOISTURE SENSITIVE

AIR SENSITIVE

SDS & Safety Documents