Please login to your online account to display your discounted pricing

Aluminium, 99.5%, shot (2-10 mm), ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  300210250

 View more versions of this product

Catalog No. AC300210250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Shot
Formula Weight 26.97
Quantity 25g
Color Gray
EINECS Number 231-072-3
TSCA TSCA
Density 2.7000g/mL
RTECS Number BD0330000
Fieser 02,19; 04,9; 12,486; 16,337
Assay Percent Range 99.5%
MDL Number MFCD00134029
CAS 7429-90-5
Packaging Glass bottle
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 AIR SENSITIVE
Total Trace Metal Impurities 0.5% max.
Chemical Name or Material Aluminium
Name Note 99.5%
Molecular Formula Al

Specifications

Chemical Name or Material Aluminium
Name Note 99.5%
CAS 7429-90-5
Assay Percent Range 99.5%
Molecular Formula Al
Formula Weight 26.97
MDL Number MFCD00134029
Accessories

MOISTURE SENSITIVE

AIR SENSITIVE

SDS & Safety Documents