Please login to your online account to display your discounted pricing

Aluminium, 99.5%, shot (2-10 mm), ACROS Organics™

$72.41 - $72.41

Chemical Identifiers

Chemical Name or Material Aluminium
Name Note 99.5%
CAS 7429-90-5
Assay Percent Range 99.5%
Molecular Formula Al
Formula Weight 26.97
MDL Number MFCD00134029
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC300210250

  SDS
thermo fisher scientific
300210250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC300212500

  SDS
thermo fisher scientific
300212500
Glass bottle 250g Each for $72.41

AC300210010

  SDS
thermo fisher scientific
AC300210010
Glass bottle 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Shot
Formula Weight 26.97
Quantity 25g
Color Gray
EINECS Number 231-072-3
TSCA TSCA
Density 2.7000g/mL
RTECS Number BD0330000
Fieser 02,19; 04,9; 12,486; 16,337
Assay Percent Range 99.5%
MDL Number MFCD00134029
CAS 7429-90-5
Packaging Glass bottle
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 AIR SENSITIVE
Total Trace Metal Impurities 0.5% max.
Chemical Name or Material Aluminium
Name Note 99.5%
Molecular Formula Al
Safety and Handling

MOISTURE SENSITIVE

AIR SENSITIVE