missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alumina DCC, MP Biomedicals™

Alumina is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories

$239.23 - $1,315.95

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminum oxide, alundum, fasertonerde
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN04512
View Documents Promotion Details
Mp Biomedicals
0204512
500g
Each for $239.23
 
ICN04511
View Documents Promotion Details
Mp Biomedicals
0204511
5kg
Each for $1,315.95
 
Description

Description

Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminum oxide, alundum, fasertonerde
dialuminium(3+) trioxidandiide

Specifications

1344-28-1
500g
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
Free Flowing Powder
3.94g/cm3 at 20°C
Pink to White
Al2O3
abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminum oxide, alundum, fasertonerde
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Solid
Alumina DCC
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Product Certifications