missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alumina DCC, MP Biomedicals™
Alumina is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories
Manufacturer: Mp Biomedicals 0204512
Specifications
Alumina DCC | |
Al2O3 | |
abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminium oxide, aluminum oxide, alundum, fasertonerde | |
[O-2].[O-2].[O-2].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
3.94g/cm3 at 20°C | |
Pink to White |
1344-28-1 | |
MFCD00003424 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
Free Flowing Powder | |
Solid | |
500g |
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O-2].[O-2].[O-2].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminium oxide, aluminum oxide, alundum, fasertonerde | |
dialuminum;oxygen(2-) |
Safety and Handling
Recommended Storage : Store at room temperature.