Please login to your online account to display your discounted pricing

Alumina C, MP Biomedicals™


Adsorbents

Quantity 1g

Manufacturer: mp biomedicals  0210025501

 View more versions of this product

Catalog No. ICN10025501

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 3.94g/cm3 at 20°C
Physical State Solid
Quantity 1g
Chemical Name or Material Alumina C
CAS 1344-28-1
Physical Form White Powder
Molecular Formula Al2O3
Formula Weight 101.93g/mol
MDL Number MFCD00003424
Synonym Aluminum oxide
Recommended Storage Store at room temperature.

Specifications

Chemical Name or Material Alumina C
CAS 1344-28-1
Physical Form White Powder
Molecular Formula Al2O3
Formula Weight 101.93g/mol
MDL Number MFCD00003424
Synonym Aluminum oxide
Accessories

Store at room temperature.