Please login to your online account to display your discounted pricing

Alumina C, MP Biomedicals™

Adsorbents

$53.45 - $53.45

Chemical Identifiers

Chemical Name or Material Alumina C
CAS 1344-28-1
Physical Form White Powder
Molecular Formula Al2O3
Formula Weight 101.93g/mol
MDL Number MFCD00003424
Synonym Aluminum oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10025501

 
mp biomedicals
0210025501
1g Each for $53.45
Description & Specifications

Specifications

Density 3.94g/cm3 at 20°C
Physical State Solid
Quantity 1g
Chemical Name or Material Alumina C
CAS 1344-28-1
Physical Form White Powder
Molecular Formula Al2O3
Formula Weight 101.93g/mol
MDL Number MFCD00003424
Synonym Aluminum oxide
Recommended Storage Store at room temperature.
Safety and Handling

Store at room temperature.