missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alumina A, Activity: Super I, MP EcoChrom™, MP Biomedicals™

Manufacturer:  Mp Biomedicals 0204592

 View more versions of this product

Catalog No. ICN04592



Description

Description

Specifications

Specifications

Alumina A
1344-28-1
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
Free Flowing Powder
200m2/g
Pink to White
Activity: Super I
Al2O3
abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminium oxide, aluminum oxide, alundum, fasertonerde
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
3.94g/cm3 at 20°C
Solid
500g

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, aluminium oxide, aluminum oxide, alundum, fasertonerde
dialuminium(3+) trioxidandiide
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

Recommended Storage : Store at room temperature.

SDS
Documents