Please login to your online account to display your discounted pricing

Altanserin hydrochloride, Tocris

5-HT2A receptor antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Altanserin hydrochloride
Assay Percent Range >98%
Molecular Formula C22H22FN3O2S·HCl
Formula Weight 447.95
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 3-dihydro-2-thioxo-4(1H)-quinazolinone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

180910

 
r & d systems
180910
10mg Each for $177.37

180950

 
r & d systems
180950
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 447.95
Quantity 10mg
Molecular Formula C22H22FN3O2S·HCl
CAS 1135280-78-2
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2, 3-dihydro-2-thioxo-4(1H)-quinazolinone hydrochloride
Chemical Name or Material Altanserin hydrochloride
Safety and Handling

Desiccate at +4°C