Please login to your online account to display your discounted pricing

Alrestatin, Tocris

Aldose reductase inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Alrestatin
Assay Percent Range >98%
Molecular Formula C14H9NO4
Formula Weight 255.23
Synonym 1, 3-Dioxo-1H-benz[d, e]isoquinoline-2(3H)-acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

048550

 
r & d systems
RDS048550
50mg Each for $682.67

048510

 
r & d systems
RDS048510
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 255.23
Chemical Name or Material Alrestatin
Molecular Formula C14H9NO4
CAS 51411-04-2
Assay Percent Range >98%
Synonym 1, 3-Dioxo-1H-benz[d, e]isoquinoline-2(3H)-acetic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature