Please login to your online account to display your discounted pricing

Alrestatin, Tocris


Aldose reductase inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS048550

 View more versions of this product

Catalog No. 048550

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 255.23
Chemical Name or Material Alrestatin
Molecular Formula C14H9NO4
Assay Percent Range >98%
CAS 51411-04-2
Synonym 1, 3-Dioxo-1H-benz[d, e]isoquinoline-2(3H)-acetic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Alrestatin
Assay Percent Range >98%
Molecular Formula C14H9NO4
Formula Weight 255.23
Synonym 1, 3-Dioxo-1H-benz[d, e]isoquinoline-2(3H)-acetic acid
Accessories

Store at Room Temperature