Please login to your online account to display your discounted pricing

Alprostadil, Tocris

Prostaglandin; Vasodilator and antiplatelet agent in vivo

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material Alprostadil
Assay Percent Range >98%
Molecular Formula C20H34O5
Formula Weight 354.49
Synonym (11α,13E,15S)-11,15-Dihydroxy-9-oxo-prost-13-en-1-oic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

162010

 
r & d systems
RDS162010
10mg Each for $208.25

162050

 
r & d systems
RDS162050
50mg Each for $867.95
Description & Specifications

Specifications

Formula Weight 354.49
Quantity 10mg
Chemical Name or Material Alprostadil
Assay Percent Range >98%
Molecular Formula C20H34O5
CAS 745-65-3
Synonym (11α,13E,15S)-11,15-Dihydroxy-9-oxo-prost-13-en-1-oic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C