Please login to your online account to display your discounted pricing

Alphaxalone, Tocris

Direct activator and potentiator of GABAA

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material Alphaxalone
Assay Percent Range >98%
Molecular Formula C21H32O3
Formula Weight 332.48
Synonym (3α, 5α)-3-Hydroxypregnane-11, 20-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

304610

 
r & d systems
RDS304610
10mg Each for $168.15

304650

 
r & d systems
RDS304650
50mg Each for $697.40
Description & Specifications

Specifications

Formula Weight 332.48
Quantity 10mg
Molecular Formula C21H32O3
Chemical Name or Material Alphaxalone
Assay Percent Range >98%
CAS 23930-19-0
Synonym (3α, 5α)-3-Hydroxypregnane-11, 20-dione
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature