missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood and Socks

Provide an extremely effective barrier against both chemical and biological hazards. Ansell™ Alphatec™ MICROCHEM™ 68-3000 Yellow Coveralls with Hood and Socks are durable and multi-layered, and are some of the most comfortable chemical protective materials available.

$341.66 - $430.43

Specifications

Latex Free No
Color Yellow
Material Barrier laminate fabric
Neck Style Hooded
Closure Type Double zipper
View More Specs

Alert:

This product contains natural rubber latex which may cause allergic reactions.


Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-000-004
View Documents
Ansell™
YE30W9212202
Small
Case of 6 Each for $341.66
 
18-000-005
View Documents
Ansell™
YE30W9212203
Medium
Case of 6 Each for $341.66
 
18-000-006
View Documents
Ansell™
YE30W9212204
Large
Case of 6 Each for $341.66
 
18-000-007
View Documents
Ansell™
YE30W9212205
X-Large
Case of 6 Each for $341.66
 
18-000-008
View Documents
Ansell™
YE30W9212206
2X-Large
Case of 6 Each for $341.66
 
18-000-009
View Documents
Ansell™
YE30W9212207
3X-Large
Case of 6 Each for $362.24
 
18-000-010
View Documents
Ansell™
YE30W9212208
4X-Large
Case of 6 Each for $391.19
 
18-000-011
View Documents
Ansell™
YE30W9212209
5X-Large
Case of 6 Each for $430.43
 
Description

Description

  • Protection: Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible: Bright yellow for improved worker safety
  • Anti-static: Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design: Elastic waist, wrists and ankles, double zip closure, double cuffs, ultrasonically welded seams, finger loops, 2 piece respirator fit hood, integrated socks with boot overflap
  • Applications: Chemicals, oil and petrochemicals, pharmaceutical, food indsutry, sewage purification installations, industrial and tank cleaning, mining
  • Sizes: Available in Small through 5X-Large
  • Certifications: EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications

Specifications

No
Barrier laminate fabric
Double zipper
Elasticated
Welded
Yellow
Hooded
Double,Elastic
Attached socks,Boot overflaps,Elastic
Liquid and spray tight chemical and biological protection
SDS