missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Collar

Provide an excellent barrier to harmful chemicals and blood borne pathogens. Ansell™ Alphatec™ MICROCHEM™ 68-2300 Yellow Coveralls with Collar are lightweight and relatively strong and durable. These coveralls are designed for applications such as environmental clean-up, sewage purification installations, and composites manufacturing.

Supplier:  Ansell™ YY23B9210305

 View more versions of this product

Catalog No. 17-999-903


Only null left
Explore available promotions

Alert:

This product contains natural rubber latex which may cause allergic reactions.


Description

Description

  • Protection:  A barrier to numerous inorganic liquid chemicals including acids and bases; achieves the highest classifications for protection from biological agents in accordance with EN 14126:2003 & ASTM F1671 for penetration of blood, bodily fluids, and blood-borne pathogens
  • Comfort:  Lightweight yet strong and durable
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, 2-way front zipper with taped storm flap, bound seams, finger loops, collar
  • Applications:  Environmental cleanup, sewage purification installations, composites manufacturing
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1674

Specifications

Specifications

X-Large
Yellow
Elastic
Collared
Bound
No
Multi-layer Barrier Laminate Fabric
Elastic
Elastic, Thumb Loop
2-way Front Zipper with Storm Flap
Particulate and light liquid splash protection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions