Please login to your online account to display your discounted pricing

AlphaProTech Critical Cover™ NaviTrak™ Shoe Covers

Designed for short wear times on non-aggressive floor surfaces.

$101.83 - $101.83

Specifications

Color Blue, White
Material NaviTrak™
Product Type Shoe Covers
For Use With (Application) Ideally used for short wear times on non-aggressive floor surfaces
Height (English) 6.25 in.
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

19-169-4134

 
alphaprotech
CR11122B
17 in. 43.18cm X-Large Case of 300 Each for $101.83
Description & Specifications

Specifications

Color Blue, White
Material NaviTrak™
Product Type Shoe Covers
For Use With (Application) Ideally used for short wear times on non-aggressive floor surfaces
Height (English) 6.25 in.
Height (Metric) 15.87cm
Seam Type Heat sealed

  • Fluid impervious
  • All-film construction
  • Heat-sealed seam delivers complete protection against fluids