Please login to your online account to display your discounted pricing

AlphaProTech Critical Cover™ GenPro™ Isolation Gowns

Lightweight and durable

$106.03 - $106.03

Specifications

Cuff Style Elastic
Color Yellow
For Use With (Application) Moderate barrier protection
Material GenPro™ Fabric
Disposable Yes
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-120-2770

 
alphaprotech
GN131244
X-Large Case of 50 Each for $106.03
Description & Specifications

Specifications

Cuff Style Elastic
Color Yellow
For Use With (Application) Moderate barrier protection
Material GenPro™ Fabric
Disposable Yes

  • For non-hazardous environments
  • Limited barrier to particulates and contamination
  • Banded ties at neck and waist