Please login to your online account to display your discounted pricing

AlphaProTech Critical Cover™ ComforTech™ Sleeves for Sterile Suits

Ultra-clean material to ensure a low particulate count and protection for sterile work areas

$463.73 - $486.93

Specifications

Cuff Style Elastic
Color White
Construction Sonically welded seams and tunneled elastic at both ends
For Use With (Equipment) Sterile suit
Sterile Yes
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

19-830-003

 
alphaprotech
MS01J768ST
19.5 in. 49.5cm Universal Case of 300 Each for $463.73

19-168-457

 
alphaprotech
MS01J764ST*
21.5 in. 54.6cm X-Large Case of 150 Pair for $486.93
Description & Specifications

Specifications

Cuff Style Elastic
Color White
Construction Sonically welded seams and tunneled elastic at both ends
For Use With (Equipment) Sterile suit
Sterile Yes

  • Sonically welded seams make it impervious to fluids, particulates and many chemicals
  • Tapered cut for secure and comfortable fit
  • Larger end of sleeve is cuffed to expose garment interior so it can be donned without touching exterior
  • Positioned inside package for immediate donning, no fumbling to properly put on sleeve
  • Sterility Assurance Level: Packaged to eliminate cross contamination, terminally sterilized using gamma irradiation to yield a minimum SAL of 10-6
  • Tunneled elastic at both ends