AlphaProTech Critical Cover™ ComforTech™ Assurance™ Coveralls
$252.36 - $315.36
Specifications
Certifications/Compliance EN 552 A1:1999, ISO 11137:1995, EN ISO 9001:2000
Closure Type Zipper front
Color White
Material Fabric
For Use With (Application) Eliminate bioburden risks to manufacturing and R & D areas with the exceptional containment performance, protects the wearer from particles and biologicals with superior particle filtration
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Size Price Quantity  
Catalog Number Mfr. No. Includes Size Price Quantity  
19-166-1190 AlphaProTech
CVAJ40224
Elastic wrist, ankle and back, inset sleeves X-Large Case of 25 Each for $252.36
19-166-1189 AlphaProTech
CVAJ40223
Elastic wrist, ankle and back, inset sleeves Large Case of 25 Each for $252.36
19-166-1191 AlphaProTech
CVAJ40225
Elastic wrist, ankle and back, inset sleeves 2X-Large Case of 25 Each for $265.09
19-166-1193 AlphaProTech
CVAJ40227
Elastic wrist, ankle and back, inset sleeves 4X-Large Case of 25 Each for $292.04
19-166-1194 AlphaProTech
CVAJ4C922
Elastic wrist, ankle and back, attached AquaTrak™ boots, inset sleeves Medium Case of 25 Each for $282.57
19-166-1192 AlphaProTech
CVAJ40226
Elastic wrist, ankle and back, inset sleeves 3X-Large Case of 25 Each for $278.12
19-166-1188 AlphaProTech
CVAJ40222
Elastic wrist, ankle and back, inset sleeves Medium Case of 25 Each for $252.36
19-166-1195 AlphaProTech
CVAJ4C923
Elastic wrist, ankle and back, attached AquaTrak™ boots, inset sleeves Large Case of 25 Each for $282.57
19-166-1198 AlphaProTech
CVAJ4C926
Elastic wrist, ankle and back, attached AquaTrak™ boots, inset sleeves 3X-Large Case of 25 Each for $315.36
19-169-4029 AlphaProTech
CVAJ4C921
Elastic wrist, ankle and back, attached AquaTrak™ boots, inset sleeves Small Case of 25 Each for $291.54
Specifications
Description
  • Terminally sterilized using gamma irradiation to yield a minimum SAL of 10-4
  • Strong, lightweight, static dissipative coveralls have an ultralow particle count
  • Exhibit a high vapor transmission rate to dispel perspiration for cool, comfortable, all-day wear
  • Barrier to fluids and particulates
  • Elastic wrist, ankle and back
  • Serged seams
  • Color: White
Description & Specifications
Specifications
EN 552 A1:1999, ISO 11137:1995, EN ISO 9001:2000
White
Eliminate bioburden risks to manufacturing and R & D areas with the exceptional containment performance, protects the wearer from particles and biologicals with superior particle filtration
Yes
Zipper front
Fabric
Yes