Please login to your online account to display your discounted pricing

AlphaProTech Critical Cover™ ChemTech™ Coveralls

Designed with a safety barrier on the inside for an added measure of protection

$567.18 - $757.59

Specifications

Color Yellow
Cuff Style Elastic
Includes Boots, elastic wrist, protective flap over zipper closure
Seam Type Bound
For Use With (Application) Chemical barrier, liquid and particle holdout
View More Specs
Products
Catalog Number Mfr. No. Length (English) Sleeve Length (Metric) Sleeve Width (English) Chest Size Price Quantity    

19-169-5037

 
alphaprotech
CVBK42127
41 in. 104.14cm 32.5 in. 4X-Large Case of 25 Each for $757.59

19-169-5035

 
alphaprotech
CVBK42125
37 in. 93.98cm 29.75 in. 2X-Large Case of 25 Each for $618.79

19-169-5032

 
alphaprotech
CVBK42122
34 in. 86.36cm 24.25 in. Medium Case of 25 Each for $567.18

19-169-5033

 
alphaprotech
CVBK42123
34 in. 86.36cm 25 in. Large Case of 25 Each for $567.18
Description & Specifications

Specifications

Color Yellow
Cuff Style Elastic
Includes Boots, elastic wrist, protective flap over zipper closure
Seam Type Bound
For Use With (Application) Chemical barrier, liquid and particle holdout

  • Chemical barrier properites
  • Liquid and particle holdout
  • Protective flap over zipper
  • Elastic wrists
  • Attached boots
  • Bound seams for additional protection

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.