AlphaProTech Critical Cover™ AquaTrak™ Shoe Covers
AquaTrak material keeps wearer stable and protected in wet and dry conditions
Manufacturer:  AlphaProTech AT31111BSL
 View more versions of this product
Catalog No. 19-130-1472
Description
Description
  • Anti-skid properties provide amazing slip and fall protection w/o additional coatings
  • Material is fluid impervious
  • Styles with heat-sealed seams provide complete protection even in standing water
  • Serged seam soles provide great fit and can be used in dry or moist conditions
  • High breathability, low particulate count fabric wears comfortably and completely contains contamination
Specifications
Specifications
Elastic top with ankle ties
Universal
Ideal for superior performance APT and amazing slip and fall protection
17.78cm
44.45cm
Black
AquaTrak™
7 in.
17.5 in.
Sonic welded
Documents