AlphaProTech Critical Cover™ AquaTrak™ Shoe Covers
AquaTrak™ material keeps the wearer stable and protected in wet and dry conditions.
Manufacturer:  AlphaProTech AT21111BSL
 View more versions of this product
Catalog No. 29-635-7901
Description
Description
  • Anti-skid properties provide amazing slip and fall protection without additional coatings
  • Material is fluid impervious
  • Styles with heat-sealed seams provide complete protection even in standing water
  • Serged seam soles provide great fit and can be used in dry or moist conditions
  • High breathability, low particulate count fabric wears comfortably and completely contains contamination
Specifications
Specifications
Elastic top with ankle ties
Universal
AquaTrak™
7 in.
17.5 in.
Sonic welded
White
Non-skid, seamless
Ideal for superior performance APT and amazing slip and fall protection
17.78cm
44.45cm