AlphaProTech Critical Cover™ AquaTrak™ Shoe Covers
AquaTrak™ material keeps the wearer stable and protected in wet and dry conditions.
Manufacturer:  AlphaProTech AT11112BSL
 View more versions of this product
Catalog No. 29-635-7900
Description
Description
  • Anti-skid properties provide amazing slip and fall protection without additional coatings
  • Material is fluid impervious
  • Styles with heat-sealed seams provide complete protection even in standing water
  • Serged seam soles provide great fit and can be used in dry or moist conditions
  • High breathability, low particulate count fabric wears comfortably and completely contains contamination
Specifications
Specifications
Elastic top with ankle ties
X-Large
AquaTrak™
7 in.
17.75 in.
Sonic welded
Blue
Non-skid, seamless
Ideal for superior performance APT and amazing slip and fall protection
17.78cm
45cm