AlphaProTech Critical Cover™ AlphaGuard™ Long Sleeve Scrub Shirts
Soft and comfortable material that provides a visible barrier and exceptional breathability
Manufacturer:  AlphaProTech WS232211
 View more versions of this product
Catalog No. 19-140-215
Description
Description
  • Shirts have round neck, banded collar, elastic wrist, one pocket, and safety stiched seams
  • Fluid repellent material that delivers high bacterial filtration rates with low particle counts reducing your contamination concerns
  • Static dissipative, perfect for applications that require an ESD material
Specifications
Specifications
22.5 in.
Safety stitched seams for added strength and durability
Safety stiched
20.5 in.
52.07cm
43.18cm
Blue
Fabric
Small
Round
1.0
Documents