AlphaProTech Critical Cover™ AlphaGuard™ Coveralls i
Fluid repellent and static dissipative
Manufacturer:  AlphaProTech CV740322
 View more versions of this product
Catalog No. 19-130-1502B
Alert:
Not recommended for use with chemicals.
Description
Description
  • Low particle count
  • Elastic back
  • Serged seams
Specifications
Specifications
Elastic
Elastic
Elastic
Suitable for use in controlled environments with low particle count
34 in.
Serged
24.25 in.
61.59 cm
AlphaGuard™ fabric
White
Elastic wrist, ankle and back
Yes
86.36 cm
Medium
28.5 in.
72.39 cm