missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
AlphaProTech UltraGrip™ BarrierTech™ Shoe Covers
The Ultimate in Slip and Fall Protection
Manufacturer: AlphaProTech SHM1315BH
Description
- Ultimate in slip and Fall Protection on damp and dry floors
- High performance, anti-skid properties
- Full coverage up through ankle area
- BarrierTech™ upper material provides protection from fluids and light chemical splash
- Two colors, four sizes
- Different colors around the opening provide convenient size identification
Specifications
Blue | |
Medium | |
Ideal for slip and fall protection on wet and dry floors | |
12.7cm | |
29.21cm | |
Yellow |
11.5 x 4.25 x 5 in. (29.21 x 10.7 x 12.7cm) | |
BarrierTech™ | |
5 in. | |
11.5 in. | |
Serged | |
Yellow |