AlphaProTech AlphaGuard™ Lab Coats
Vapor transmission for cool and comfortable wear
Manufacturer:  AlphaProTech LC326326
 View more versions of this product
Catalog No. 19-130-1466F
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
Vapor transmission for cool and comfortable wear
  • Soft, breatheable and fluid repellent
  • Static dissipative
  • Ultra clean, low particle count
Specifications
Specifications
Tapered
White
AlphaGuard™ fabric
3X-Large
97.1cm
30.75 in.
113.03 cm
Snaps
Knit
3
Yes
Serged
78.1 cm
44.5 in.