Learn More
alpha-Toluenesulfonyl chloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1012422
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
alpha-Toluenesulfonyl chloride | |
89°C to 94°C | |
C7H7ClO2S | |
100 g | |
972806 | |
alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride | |
C1=CC=C(C=C1)CS(=O)(=O)Cl | |
190.641 | |
190.65 |
1939-99-7 | |
120°C (10 mmHg) | |
MFCD00007455 | |
UN3096 | |
Moisture sensitive | |
OAHKWDDSKCRNFE-UHFFFAOYSA-N | |
phenylmethanesulfonyl chloride | |
74740 | |
99% |
Chemical Identifiers
1939-99-7 | |
190.641 | |
OAHKWDDSKCRNFE-UHFFFAOYSA-N | |
74740 | |
C1=CC=C(C=C1)CS(=O)(=O)Cl |
C7H7ClO2S | |
MFCD00007455 | |
alpha-toluenesulfonyl chloride, benzylsulfonyl chloride, benzenemethanesulfonyl chloride, benzylsulfochloride, .alpha.-toluenesulfonyl chloride, methanesulfonyl chloride, phenyl, alpha-toluenesulfonylchloride, phenylmethylsulfonyl chloride, phenylmethanesulphonyl chloride, phenyl-methanesulfonyl chloride | |
phenylmethanesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 217-717-1
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only