Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Terpinyl acetate, tech. 90%

$49.27 - $167.26

Chemical Identifiers

Chemical Name or Material alpha-Terpinyl acetate
CAS 80-26-2
Purity Grade Technical Grade
Molecular Formula C12H20O2
Formula Weight 196.28
MDL Number MFCD00037155
Synonym alpha,alpha,4-Trimethyl-3-cyclohexene-1-methanol acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1795222

  SDS
alfa aesar™
A1795222
100g Each for $49.27

AAA1795236

  SDS
alfa aesar™
A1795236
500g Each for $167.26
Description & Specifications

Specifications

Formula Weight 196.28
Chemical Name or Material alpha-Terpinyl acetate
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
Molecular Formula C12H20O2
MDL Number MFCD00037155
CAS 80-26-2
Synonym alpha,alpha,4-Trimethyl-3-cyclohexene-1-methanol acetate
Health Hazard 1 Warning
Boiling Point 220°C
Quantity 100g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 99°C (210°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number OT0200000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.