missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-Terpineol, 96%, Thermo Scientific Chemicals

A naturally-occuring monoterpene alcohol

Supplier:  Thermo Scientific Chemicals 01628536

 View more versions of this product

Catalog No. AA1628536



Description

Description

A naturally-occuring monoterpene alcohol.Alpha-terpineol is used as an antioxidant, antiseptic, antihypernociception and anti-inflammatory. It is also used as a solvent. It is an important ingredient of pine oil disinfectants. Further, it is used as a fragrance in perfumes, fat denaturant for soap production and synthetic flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

alpha-Terpineol
31°C to 32°C
0.930 g/mL
90°C (194°F)
1.483
500 g
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
WUOACPNHFRMFPN-UHFFFAOYSA-N
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
17100
154.24
Liquid
98-55-5
Colorless to Yellow
210°C to 219°C
C10H18O
MFCD00001557
14,9171
Miscible with water,benzene,propylene glycol,acetone,alcohol and ether.
CC1=CCC(CC1)C(C)(C)O
154.253
CHEBI:22469
96%

Chemical Identifiers

98-55-5
154.253
WUOACPNHFRMFPN-UHFFFAOYSA-N
17100
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
C10H18O
MFCD00001557
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
CHEBI:22469
CC1=CCC(CC1)C(C)(C)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 202-680-6

RTECSNumber : WZ6700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only