missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Terpineol, 96%

A naturally-occuring monoterpene alcohol

Manufacturer:  Alfa Aesar™ 1628522

 View more versions of this product

Catalog No. AA1628522



Specifications

Specifications

alpha-Terpineol
0.930g/mL
C10H18O
14,9171
WUOACPNHFRMFPN-UHFFFAOYSA-N
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
17100
1.4830
Liquid
Colorless to Yellow
90° (194°F)
98-55-5
96%
MFCD00001557
alpha,alpha,4-Trimethyl-3-cyclohexene-1-methanol; p-Menth-1-en-8-ol
CC1=CCC(CC1)C(C)(C)O
154.253
CHEBI:22469
154.24
210°C to 219°C
31°C to 32°C
100g

Chemical Identifiers

98-55-5
154.253
WUOACPNHFRMFPN-UHFFFAOYSA-N
17100
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
C10H18O
MFCD00001557
alpha,alpha,4-Trimethyl-3-cyclohexene-1-methanol; p-Menth-1-en-8-ol
CHEBI:22469
CC1=CCC(CC1)C(C)(C)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H227
Causes skin irritation.
Combustible liquid.

GHS P Statement
P210-P280-P362-P321-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WZ6700000

TSCA : TSCA

SDS Documents