missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alpha-Smooth Muscle Actin, eFluor 570, clone: 1A4, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS41976082

 View more versions of this product

Catalog No. 501130701



Description

Description

Among the six actin isoforms described in mammals, two are found in virtually all cells (b- and g-cytoplasmic), two are detected in smooth muscle cells (a- and g-smooth muscle) and two are present in striated muscles, one predominantly in skeletal (a-skeletal) and one in cardiac (a-cardiac) muscle cells. These actin isoforms differ slightly in their N-terminus and the sequences of each of them are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown to appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases.

Description: The monoclonal antibody 1A4 recognizes human, mouse, and rat alpha-smooth muscle actin. Alpha-smooth muscle actin is a 42kDa protein and is a major component of the cytoskeletal structural network. This 1A4 antibody is specific for the alpha form of muscle actin only, which is expressed by smooth muscle cells of blood vessels, myofibroblasts, and myoepithelial cells. It is also expressed in the parenchyma and stroma of various tissues. Alpha-smooth muscle actin is useful in the identification of leiomyomas, leiomycosarcomas and pleomorphic adenomas and is used as a prognostic marker for basal cell carcinoma.Applications Reported: This 1A4 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.Applications Tested: This 1A4 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable.Excitation: 555 nm; Emission: 570 nm
Specifications

Specifications

Alpha-Smooth Muscle Actin
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P62738, P62737, P62736
Actin, aortic smooth muscle, Alpha Actin 2, alpha-sma
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Murine, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
1A4
eFluor 570
ACTA2
Liquid
Acta2
IgG2a, kappa, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
11475, 59, 81633
Documents

For Research Use Only.