Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-(Phenylthio)phenylacetic acid, 99%

$53.49 - $53.49

Chemical Identifiers

Chemical Name or Material alpha-(Phenylthio)phenylacetic acid
CAS 10490-07-0
Molecular Formula C14H12O2S
Formula Weight 244.31
MDL Number MFCD00066259
Beilstein 2373923
Synonym 2-(Phenylthio)phenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1622614

  SDS
alfa aesar™
L1622614
25g Each for $53.49
Description & Specifications

Specifications

Formula Weight 244.31
Chemical Name or Material alpha-(Phenylthio)phenylacetic acid
Molecular Formula C14H12O2S
MDL Number MFCD00066259
CAS 10490-07-0
Assay 99%
Beilstein 2373923
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2-(Phenylthio)phenylacetic acid
Melting Point 99° to 101°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.