Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Phenylglycinonitrile hydrochloride, tech. 85%

Chemical Identifiers

Chemical Name or Material alpha-Phenylglycinonitrile hydrochloride
CAS 53641-60-4
Purity Grade Technical Grade
Molecular Formula C8H8N2·HCl
Formula Weight 168.63
MDL Number MFCD00013022
UN Number UN3439
Beilstein 4155088
Synonym alpha-Aminobenzyl cyanide hydrochloride; alpha-Aminophenylacetonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0635506

 
alfa aesar™
L0635506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.63
Chemical Name or Material alpha-Phenylglycinonitrile hydrochloride
Assay 85%
Molecular Formula C8H8N2·HCl
MDL Number MFCD00013022
CAS 53641-60-4
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade
UN Number UN3439
Beilstein 4155088
Synonym alpha-Aminobenzyl cyanide hydrochloride; alpha-Aminophenylacetonitrile hydrochloride
Health Hazard 1 Danger
Melting Point ≈164°C
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
RTECS Number AL7820000
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.