missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Phenylcyclopentylacetic acid, 98%

$54.46 - $178.01

Chemical Identifiers

CAS 3900-93-4
Molecular Formula C13H16O2
Formula Weight 204.27
MDL Number MFCD00001379
Beilstein 1956835
Synonym alpha-Cyclopentylphenylacetic acid
Chemical Name or Material alpha-Phenylcyclopentylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0795506 SDS Alfa Aesar™
L0795506
5g Each for $54.46
AAL0795514 SDS Alfa Aesar™
L0795514
25g Each for $178.01
Specifications

Description

Applications
alpha-Phenylcyclopentylacetic acid is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool and dark place. Store away from incompatible materials such as oxidizing agents.
Description & Specifications

Chemical Identifiers

3900-93-4
204.27
1956835
alpha-Phenylcyclopentylacetic acid
C13H16O2
MFCD00001379
alpha-Cyclopentylphenylacetic acid

Specifications

3900-93-4
204.27
1956835
98° to 102°C
alpha-Phenylcyclopentylacetic acid
C13H16O2
MFCD00001379
alpha-Cyclopentylphenylacetic acid
5g
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA