Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-p-Xylenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material alpha-p-Xylenesulfonyl chloride
CAS 51419-59-1
Molecular Formula C8H9ClO2S
Formula Weight 204.67
MDL Number MFCD01631931
UN Number UN3261
Synonym 4-Methylbenzylsulfonyl chloride; p-Tolylmethanesulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2603803

  SDS
alfa aesar™
H2603803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.67
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 97%
Health Hazard 1 Danger
Melting Point 75° to 81°C
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3261
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Molecular Formula C8H9ClO2S
Synonym 4-Methylbenzylsulfonyl chloride; p-Tolylmethanesulfonyl chloride
CAS 51419-59-1
Chemical Name or Material alpha-p-Xylenesulfonyl chloride
MDL Number MFCD01631931
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.