missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

alpha-(p-Toluenesulfonyl)-4-fluorobenzylisonitrile, 97%, Thermo Scientific™

$122.00 - $397.52

Chemical Identifiers

CAS 165806-95-1
Molecular Formula C15H12FNO2S
Molecular Weight (g/mol) 289.32
MDL Number MFCD03787932
InChI Key UXCQPEDHCCJBNL-UHFFFAOYNA-N
Synonym a-tosyl-4-fluorobenzyl isocyanide, 1-fluoro-4-isocyano tosyl methyl benzene, alpha-p-toluenesulfonyl-4-fluorobenzylisonitrile, a-p-toluenesulfonyl-4-fluorobenzylisonitrile, 4-fluorophenyl isocyano methyl 4-methylphenyl sulphone, alpha-4-methylphenyl sulphonyl-4-fluorobenzylisonitrile, alpha-tosyl-4-fluorobenzyl isocyanide, alpha-p-toluenesulfonyl-4-fluorobezylisonitrile, 1-4-fluorophenyl-1-tosyl methyl isocyanide
PubChem CID 10827289
IUPAC Name 1-[(4-fluorophenyl)-isocyanomethyl]sulfonyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C([N+]#[C-])C1=CC=C(F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH52580MD
View Documents Promotion Details
Thermo Scientific Chemicals
H52580MD
250 mg
Each for $122.00
 
AAH5258003
View Documents Promotion Details
Thermo Scientific Chemicals
H5258003
1 g
Each for $397.52
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

165806-95-1
289.32
UXCQPEDHCCJBNL-UHFFFAOYNA-N
10827289
CC1=CC=C(C=C1)S(=O)(=O)C([N+]#[C-])C1=CC=C(F)C=C1
C15H12FNO2S
MFCD03787932
a-tosyl-4-fluorobenzyl isocyanide, 1-fluoro-4-isocyano tosyl methyl benzene, alpha-p-toluenesulfonyl-4-fluorobenzylisonitrile, a-p-toluenesulfonyl-4-fluorobenzylisonitrile, 4-fluorophenyl isocyano methyl 4-methylphenyl sulphone, alpha-4-methylphenyl sulphonyl-4-fluorobenzylisonitrile, alpha-tosyl-4-fluorobenzyl isocyanide, alpha-p-toluenesulfonyl-4-fluorobezylisonitrile, 1-4-fluorophenyl-1-tosyl methyl isocyanide
1-[(4-fluorophenyl)-isocyanomethyl]sulfonyl-4-methylbenzene

Specifications

165806-95-1
MFCD03787932
a-tosyl-4-fluorobenzyl isocyanide, 1-fluoro-4-isocyano tosyl methyl benzene, alpha-p-toluenesulfonyl-4-fluorobenzylisonitrile, a-p-toluenesulfonyl-4-fluorobenzylisonitrile, 4-fluorophenyl isocyano methyl 4-methylphenyl sulphone, alpha-4-methylphenyl sulphonyl-4-fluorobenzylisonitrile, alpha-tosyl-4-fluorobenzyl isocyanide, alpha-p-toluenesulfonyl-4-fluorobezylisonitrile, 1-4-fluorophenyl-1-tosyl methyl isocyanide
CC1=CC=C(C=C1)S(=O)(=O)C([N+]#[C-])C1=CC=C(F)C=C1
289.32
289.32
alpha-(p-Toluenesulfonyl)-4-fluorobenzylisonitrile
C15H12FNO2S
250 mg
UXCQPEDHCCJBNL-UHFFFAOYNA-N
1-[(4-fluorophenyl)-isocyanomethyl]sulfonyl-4-methylbenzene
10827289
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'