Please login to your online account to display your discounted pricing

alpha-Naphthoflavone, 99.5%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128160010

 View more versions of this product

Catalog No. AC128160010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Physical Form Crystalline Powder or Crystals
Formula Weight 272.29
Quantity 1g
Color Off-white to light yellow
EINECS Number 210-071-1
TSCA TSCA
RTECS Number QL6250000
Melting Point 155° to 158°C
Beilstein 17, IV, 5551
CAS Max % 100.0
Packaging Glass bottle
Synonym 7, 8-Benzoflavone
CAS Min % 99.5
Infrared Spectrum Authentic
Chemical Name or Material α-Naphthoflavone
Name Note 99.5+%
Assay Percent Range 99.5%
Molecular Formula C19H12O2
MDL Number MFCD00004985
CAS 604-59-1

Specifications

Chemical Name or Material α-Naphthoflavone
Name Note 99.5+%
CAS 604-59-1
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5%
Molecular Formula C19H12O2
Formula Weight 272.29
MDL Number MFCD00004985
Synonym 7, 8-Benzoflavone