Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Naphthoflavone, 97%

$76.99 - $76.99

Chemical Identifiers

Chemical Name or Material alpha-Naphthoflavone
CAS 604-59-1
Molecular Formula C19H12O2
Formula Weight 272.31
MDL Number MFCD00004985
Beilstein 210862
Synonym 7,8-Benzoflavone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1854206

  SDS
alfa aesar™
A1854206
5g Each for $76.99
Description & Specifications

Specifications

Formula Weight 272.31
Odor Odorless
Melting Point 156° to 159°C
MDL Number MFCD00004985
CAS 604-59-1
RTECS Number QL6250000
Synonym 7,8-Benzoflavone
Molecular Formula C19H12O2
Quantity 5g
Chemical Name or Material alpha-Naphthoflavone
TSCA TSCA
Assay 97%
Beilstein 210862