Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Methylbenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Chemical Identifiers

Chemical Name or Material alpha-Methylbenzylzinc chloride
CAS 85459-20-7
Formula Weight 250.99
MDL Number MFCD01311436
UN Number UN3399
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58185AD

  SDS
alfa aesar™
H58185AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.99
Density 0.965g/mL
Color Yellow to brown to black
Concentration 0.5M in THF
CAS 85459-20-7
UN Number UN3399
Health Hazard 2 GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Physical Form Liquid
Storage Note 1 2° to 8°C
Packaging ChemSeal™ bottles
MDL Number MFCD01311436
DOT Information Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Chemical Name or Material alpha-Methylbenzylzinc chloride
Health Hazard 1 Danger
Quantity 50mL
Sensitivity Air Sensitive
Health Hazard 3 GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It helps in the synyhesis of of heterotactic polylactic acid from rac-lactide.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and water.

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

2° to 8°C