Please login to your online account to display your discounted pricing

Alfa Aesar™ alpha-Methyl-DL-phenylalanine, 98%

$77.73 - $745.01

Chemical Identifiers

Chemical Name or Material alpha-Methyl-DL-phenylalanine
CAS 1132-26-9
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00010512
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2515501

 
alfa aesar™
B2515501
0.25g Each for $77.73

AAB2515506

 
alfa aesar™
B2515506
5g Each for $745.01
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material alpha-Methyl-DL-phenylalanine
Assay 98%
Molecular Formula C10H13NO2
MDL Number MFCD00010512
CAS 1132-26-9
Melting Point 293° to 294°C (dec.)
Quantity 0.25g