Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, 98%

Chemical Identifiers

Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
CAS 64312-89-6
Molecular Formula C10H8ClF3O2
Formula Weight 252.62
MDL Number MFCD00078209
UN Number UN3265
Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2584503

  SDS
alfa aesar™
H2584503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2584506

  SDS
alfa aesar™
H2584506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH25845MD

  SDS
alfa aesar™
H25845MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.62
Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Assay 98%
Molecular Formula C10H8ClF3O2
MDL Number MFCD00078209
CAS 64312-89-6
UN Number UN3265
Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionyl chloride
Health Hazard 1 Danger
Boiling Point 213° to 214°C
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Flash Point 89°C (192°F)
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

2° to 8°C