Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile
CAS 80866-87-1
Molecular Formula C10H8F3NO
Formula Weight 215.18
MDL Number MFCD00001844
UN Number UN3276
Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2584406

 
alfa aesar™
H2584406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2584414

 
alfa aesar™
H2584414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.18
Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile
Assay 98%
Molecular Formula C10H8F3NO
MDL Number MFCD00001844
CAS 80866-87-1
UN Number UN3276
Synonym 3,3,3-Trifluoro-2-methoxy-2-phenylpropionitrile
Health Hazard 1 Warning
Boiling Point 76° to 82°C (12mmHg)
Quantity 5g
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Flash Point 89°C (152°F)
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H302-H312-H332-H315-H319-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.