Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 97%

$83.58 - $83.58

Chemical Identifiers

Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid
CAS 81655-41-6
Molecular Formula C10H9F3O3
Formula Weight 234.18
MDL Number MFCD00066637
Beilstein 2215439
Synonym (+/-)-Mosher's acid; (+/-)-MTPA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1435803

 
alfa aesar™
L1435803
1g Each for $83.58
Description & Specifications

Specifications

Formula Weight 234.18
Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid
CAS 81655-41-6
Assay 97%
Molecular Formula C10H9F3O3
MDL Number MFCD00066637
Beilstein 2215439
Health Hazard 1 Warning
Melting Point 40° to 44°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Synonym (+/-)-Mosher's acid; (+/-)-MTPA
Boiling Point 105° to 107°C (1mmHg)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C